2-(2-Methyl-2H-tetrazol-5-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%
Supplier: Ambeed
|
Danger
|
A134966-250MG
A134966-5G
77137-052EA
8.49
USD
77137-052
77137-054
2-(2-Methyl-2H-tetrazol-5-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%
2-(2-Methyl-2H-tetrazol-5-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BN₅O₂ MW: 287.12 g/mol |
MDL Number:
MFCD18382903 CAS Number: 1056039-83-8 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...