(R)-3-Benzylmorpholine 95%

Supplier: Ambeed
Warning

A134410-100MG A134410-1G
77137-036EA 41.38 USD
77137-036 77137-038
(R)-3-Benzylmorpholine 95%
(R)-3-Benzylmorpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.24 g/mol
MDL Number: MFCD11519114
CAS Number: 481038-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR