2-Fluoro-3-(methylthio)pyridine 95%

Supplier: Ambeed
Warning

A134338-1G A134338-250MG A134338-25G A134338-5G
77137-020EA 19.57 USD
77137-020 77137-022 77137-024 77137-026
2-Fluoro-3-(methylthio)pyridine 95%
2-Fluoro-3-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₆FNS
MW: 143.182 g/mol
MDL Number: MFCD22543684
CAS Number: 210992-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR