Dimethyl [2,2'-bipyridine]-3,3'-dicarboxylate 97%

Supplier: Ambeed
Warning

A134184-100MG A134184-10G A134184-250MG A134184-5G
77136-998EA 9.27 USD
77136-998 77137-000 77137-002 77137-004
Dimethyl [2,2'-bipyridine]-3,3'-dicarboxylate 97%
Dimethyl [2,2'-bipyridine]-3,3'-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₄
MW: 272.26 g/mol
MDL Number: MFCD00226069
CAS Number: 39775-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR