6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine 97%

Supplier: Ambeed
Danger

A133420-100MG A133420-1G
77136-980EA 62.83 USD
77136-980 77136-982
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine 97%
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₃ClF₃N₃
MDL Number: MFCD28677466
CAS Number: 1431720-68-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR