(4-Chloro-3-(pyridin-3-ylmethoxy)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A133381-1G A133381-250MG A133381-5G
77136-970EA 156.56 USD
77136-970 77136-972 77136-974
(4-Chloro-3-(pyridin-3-ylmethoxy)phenyl)boronic acid 97%
(4-Chloro-3-(pyridin-3-ylmethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BClNO₃
MDL Number: MFCD18837608
CAS Number: 2096341-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR