Thiazol-2-ylmethanol 95%

Supplier: Ambeed
Warning

A133062-100MG A133062-1G A133062-250MG A133062-5G
77136-962EA 8.49 USD
77136-962 77136-964 77136-966 77136-968
Thiazol-2-ylmethanol 95%
Thiazol-2-ylmethanol

Precaución: for Research Use Only

Formula: C₄H₅NOS
MW: 115.1500 g/mol
MDL Number: MFCD06200855
CAS Number: 14542-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR