Z-Asp(OtBu)-OH·H2O 97%

Supplier: Ambeed
Warning

A132478-100G A132478-25G A132478-5G
77136-946EA 114.33 USD
77136-946 77136-948 77136-950
Z-Asp(OtBu)-OH·H2O 97%
Z-Asp(OtBu)-OH·H2O

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆·H₂O
MDL Number: MFCD00150667
CAS Number: 229957-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR