7-Bromo-3-nitroquinolin-4-amine 95%

Supplier: Ambeed
Warning

A131565-100MG A131565-250MG
77136-922EA 93.36 USD
77136-922 77136-924
7-Bromo-3-nitroquinolin-4-amine 95%
7-Bromo-3-nitroquinolin-4-amine

Precaución: for Research Use Only

Formula: C₉H₆BrN₃O₂
MW: 268.07 g/mol
MDL Number: MFCD25956853
CAS Number: 1232149-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR