1-Iodo-3-(trifluoromethoxy)benzene 97%

Supplier: Ambeed
Warning

A130922-10G A130922-1G A130922-5G A130922-25G
77136-910EA 23.68 USD
77136-910 77136-912 77136-914 77685-139
1-Iodo-3-(trifluoromethoxy)benzene 97%
1-Iodo-3-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄F₃IO
MDL Number: MFCD01090992
CAS Number: 198206-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR