3-(2-Bromophenyl)pyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A130660-100MG A130660-250MG
77136-902EA 166.86 USD
77136-902 77136-904
3-(2-Bromophenyl)pyrrolidine hydrochloride 97%
3-(2-Bromophenyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrClN
MDL Number: MFCD11223592
CAS Number: 1203682-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR