(S)-4-Fluoro-5-((2-methyl-1,4-diazepan-1-yl)sulfonyl)isoquinoline hydrochloride dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: Ripasudil

A130094-10MG A130094-25MG A130094-1G A130094-1MG A130094-250MG A130094-2MG A130094-5MG A130094-100MG A130094-50MG
77136-882EA 31.83 USD
77136-882 77136-884 77615-016 77615-018 77615-020 77615-022 77615-024 77685-127 77685-128
(S)-4-Fluoro-5-((2-methyl-1,4-diazepan-1-yl)sulfonyl)isoquinoline hydrochloride dihydrate 98%
(S)-4-Fluoro-5-((2-methyl-1,4-diazepan-1-yl)sulfonyl)isoquinoline hydrochloride dihydrate

Precaución: for Research Use Only

Formula: C₁₅H₂₃ClFN₃O₄S
MW: 395.88 g/mol
MDL Number: MFCD26960897
CAS Number: 887375-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR