tert-Butyl 3-amino-4-cyano-2,5-dihydro-1H-pyrrole-1-carboxylate 97%

Supplier: Ambeed
Warning

A129520-100MG A129520-250MG
77136-858EA 123.07 USD
77136-858 77136-860
tert-Butyl 3-amino-4-cyano-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
tert-Butyl 3-amino-4-cyano-2,5-dihydro-1H-pyrrole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₂
MW: 209.24 g/mol
MDL Number: MFCD21602487
CAS Number: 1227461-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR