4-Chloro-7-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

A129434-100MG A129434-1G A129434-250MG
77136-852EA 11.33 USD
77136-852 77136-854 77136-856
4-Chloro-7-methyl-1H-indazole 95%
4-Chloro-7-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MDL Number: MFCD23705392
CAS Number: 1427430-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR