(2-Bromo-thiophen-3-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: (2-Bromothiophen-3-yl)methanol

A129093-250MG A129093-1G A129093-5G
77136-850EA 38.2 USD
77136-850 77685-116 77685-117
(2-Bromo-thiophen-3-yl)methanol 97%
(2-Bromo-thiophen-3-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅BrOS
MW: 193.062 g/mol
MDL Number: MFCD09025881
CAS Number: 70260-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR