WS6 98%

Supplier: Ambeed
Warning

Synonyms: N-(6-(4-(2-((4-methylpiperazin-1yl)methyl)-3-(trifluoromethyl)phenyl)amino)-2-oxoethyl)phenoxy)pyrimidin-4-yl)cyclopropanecarboxamide, 4-[[6-[(Cyclopropylcarbonyl)amino]-4-pyrimidinyl]oxy]-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-benzeneacetamide, N-(6-(4-(2-((4-((4-Methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)amino)-2-oxoethyl)phenoxy)pyrimidin-4-yl)cyclopropanecarboxamide

A129008-10MG A129008-1MG A129008-50MG A129008-5MG
77136-842EA 48.81 USD
77136-842 77136-844 77136-846 77136-848
WS6 98%
WS6

Precaución: for Research Use Only

Formula: C₂₉H₃₁F₃N₆O₃
MW: 568.59 g/mol
MDL Number: MFCD26793882
CAS Number: 1421227-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR