(3S,6S)-2,7-Dimethyloctane-3,6-diol 98%

Supplier: Ambeed
Warning

A128426-100MG A128426-1G A128426-250MG
77136-828EA 42.44 USD
77136-828 77136-830 77136-832
(3S,6S)-2,7-Dimethyloctane-3,6-diol 98%
(3S,6S)-2,7-Dimethyloctane-3,6-diol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₂
MW: 174.28 g/mol
MDL Number: MFCD01631077
CAS Number: 129705-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR