(5-Bromo-2-chlorophenyl)(4-ethoxyphenyl)methanone 97%

Supplier: Ambeed
Warning

A127840-100G A127840-10G A127840-1G A127840-250MG A127840-25G A127840-5G
77136-802EA 146.41 USD
77136-802 77136-804 77136-806 77136-808 77136-810 77136-812
(5-Bromo-2-chlorophenyl)(4-ethoxyphenyl)methanone 97%
(5-Bromo-2-chlorophenyl)(4-ethoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrClO₂
MW: 339.61 g/mol
MDL Number: MFCD11044417
CAS Number: 461432-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR