2-Chloro-6-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-chloro-α,α,α-trifluortoluene, 2-Amino-3-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-o-toluidine, 2-Amino-3-chlorobenzotrifluoride, 2-Chloro-6-trifluoromethylaniline

A127637-100G A127637-10G A127637-1G A127637-25G A127637-5G
77136-788EA 111.24 USD
77136-788 77136-790 77136-792 77136-794 77136-796
2-Chloro-6-(trifluoromethyl)aniline 98%
2-Chloro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MDL Number: MFCD00272565
CAS Number: 433-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR