4-Chloro-2-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-methoxy-3-nitrobenzene

A127151-100G A127151-25G A127151-5G
77136-772EA 57.68 USD
77136-772 77136-774 77136-776
4-Chloro-2-nitroanisole 98%
4-Chloro-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD00024327
CAS Number: 89-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR