3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-5-hydroxy-6,8-dimethylpyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione 98%

Supplier: Ambeed
Warning

A127099-100MG A127099-1G A127099-250MG A127099-5G
77136-766EA 13.8 USD
77136-766 77136-768 77136-770 77566-462
3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-5-hydroxy-6,8-dimethylpyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione 98%
3-Cyclopropyl-1-(2-fluoro-4-iodophenyl)-5-hydroxy-6,8-dimethylpyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione

Precaución: For research use only.

Formula: C₁₈H₁₅FIN₃O₄
MW: 483.23 g/mol
MDL Number: MFCD18207185
CAS Number: 871700-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR