(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sitagliptin
A127061-100MG
A127061-1G
A127061-5G
A127061-100G
A127061-10G
A127061-25G
77136-760EA
8.49
USD
77136-760
77136-762
77136-764
77745-921
77745-922
77745-923
(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one 98%
(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one
Precaución: for Research Use Only
Formula:
C₁₆H₁₅F₆N₅O MW: 407.31 g/mol |
MDL Number:
MFCD19684081 CAS Number: 486460-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...