Methyl 3,5-difluoro-4-formylbenzoate 97%
Supplier: Ambeed
|
Warning
|
A127000-100MG
A127000-10G
A127000-1G
A127000-250MG
A127000-5G
77136-750EA
54.11
USD
77136-750
77136-752
77136-754
77136-756
77136-758
Methyl 3,5-difluoro-4-formylbenzoate 97%
Methyl 3,5-difluoro-4-formylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₆F₂O₃ MW: 200.139 g/mol |
MDL Number:
MFCD28525711 CAS Number: 1415124-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...