(2,6-Dichlorobenzyl)methylamine hydrochloride 97%

Supplier: Ambeed
Warning

A126980-1G A126980-250MG A126980-5G
77136-744EA 32.96 USD
77136-744 77136-746 77136-748
(2,6-Dichlorobenzyl)methylamine hydrochloride 97%
(2,6-Dichlorobenzyl)methylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀Cl₃N
MDL Number: MFCD05864732
CAS Number: 90389-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR