(2-(Butylthio)pyrimidin-5-yl)boronic acid 97%

Supplier: Ambeed
Warning

A126412-100MG A126412-1G A126412-250MG
77136-736EA 36.08 USD
77136-736 77136-738 77136-740
(2-(Butylthio)pyrimidin-5-yl)boronic acid 97%
(2-(Butylthio)pyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₃BN₂O₂S
MW: 212.07 g/mol
MDL Number: MFCD18434409
CAS Number: 2096333-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR