(Fmoc-Cys-OtBu)₂ 97%

Supplier: Ambeed
Warning

Synonyms: (Fmoc-Cys-Otbu)2

A126254-100G A126254-10G A126254-1G A126254-250MG A126254-25G
77136-726EA 815.84 USD
77136-726 77136-728 77136-730 77136-732 77136-734
(Fmoc-Cys-OtBu)₂ 97%
(Fmoc-Cys-OtBu)₂

Precaución: for Research Use Only

Formula: C₄₄H₄₈N₂O₈S₂
MW: 797.01 g/mol
MDL Number: MFCD08458535
CAS Number: 139592-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR