1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
[(2,4-dichlorophenyl)methyl](methyl)amine hydrochloride, N-Methyl-2,4-dichlorobenzylamine Hydrochloride
A125894-100G
A125894-10G
A125894-1G
A125894-25G
77136-714EA
186.72
USD
77136-714
77136-716
77136-718
77136-720
1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride 97%
1-(2,4-Dichlorophenyl)-N-methylmethanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀Cl₃N MW: 226.5300 g/mol |
MDL Number:
MFCD04117880 CAS Number: 90389-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...