N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)-2-nitrobenzene-1,4-diamine 95%

Supplier: Ambeed
Warning

A125637-10G A125637-1G A125637-250MG A125637-25G A125637-5G
77136-698EA 165.51 USD
77136-698 77136-700 77136-702 77136-704 77136-706
N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)-2-nitrobenzene-1,4-diamine 95%
N1-(2-(Dimethylamino)ethyl)-5-methoxy-N1-methyl-N4-(4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)-2-nitrobenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₂₅H₂₉N₇O₃
MW: 475.54 g/mol
MDL Number: MFCD28167947
CAS Number: 1421372-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR