5-Fluoro-4-iodopyridin-2-amine 98%

Supplier: Ambeed
Warning

A125314-1G A125314-250MG A125314-25G A125314-5G
77136-680EA 16.48 USD
77136-680 77136-682 77549-482 77566-412
5-Fluoro-4-iodopyridin-2-amine 98%
5-Fluoro-4-iodopyridin-2-amine

Precaución: For research use only.

Formula: C₅H₄FIN₂
MDL Number: MFCD28722380
CAS Number: 1649470-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR