Ethyl 3-(4-chlorophenyl)propiolate 97%

Supplier: Ambeed
Warning

A125204-1G A125204-250MG A125204-5G
77136-674EA 31.94 USD
77136-674 77136-676 77136-678
Ethyl 3-(4-chlorophenyl)propiolate 97%
Ethyl 3-(4-chlorophenyl)propiolate

Precaución: for Research Use Only

Formula: C₁₁H₉ClO₂
MDL Number: MFCD00456510
CAS Number: 20026-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR