(2-Carboxyethyl)dimethylsulfonium chloride 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl-beta-propiothetin

A125183-1000G A125183-100G A125183-10G A125183-25G A125183-500G
77136-664EA 72.1 USD
77136-664 77136-666 77136-668 77136-670 77136-672
(2-Carboxyethyl)dimethylsulfonium chloride 97%
(2-Carboxyethyl)dimethylsulfonium chloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClO₂S
MW: 170.65 g/mol
MDL Number: MFCD00142888
CAS Number: 4337-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR