3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: mCBP

A124891-1G A124891-250MG
77136-660EA 108.22 USD
77136-660 77136-662
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl 97%
3,3'-Di(9H-carbazol-9-yl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₆H₂₄N₂
MW: 484.60 g/mol
MDL Number: MFCD28138084
CAS Number: 342638-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR