Fmoc-trans-4-hydroxy-D-proline 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-trans-D-Hyp-OH, (2R,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid

A124697-1G A124697-5G
77136-658EA 40.17 USD
77136-658 77571-486
Fmoc-trans-4-hydroxy-D-proline 95%
Fmoc-trans-4-hydroxy-D-proline

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₅
MDL Number: MFCD01862677
CAS Number: 139262-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR