(3R,5S)-5-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

A124379-10G A124379-1G A124379-250MG A124379-25G A124379-5G
77136-648EA 72.15 USD
77136-648 77136-650 77136-652 77136-654 77136-656
(3R,5S)-5-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride 97%
(3R,5S)-5-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.60 g/mol
MDL Number: MFCD09056830
CAS Number: 478922-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR