(R)-3-Amino-2-hydroxypropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-D-IsoSer-OH

A123417-100MG A123417-250MG A123417-1G A123417-5G
77136-606EA 82.4 USD
77136-606 77136-608 77592-408 77745-905
(R)-3-Amino-2-hydroxypropanoic acid 95%
(R)-3-Amino-2-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₃H₇NO₃
MW: 105.09 g/mol
MDL Number: MFCD05863991
CAS Number: 632-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR