3-Benzyloxyphenylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-(3-(Benzyloxy)phenyl)acetic acid

A122724-100MG A122724-1G A122724-250MG A122724-5G
77136-582EA 13.8 USD
77136-582 77136-584 77136-586 77136-588
3-Benzyloxyphenylacetic acid 98%
3-Benzyloxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
MDL Number: MFCD02664806
CAS Number: 1860-58-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR