(2-Cyanophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A122548-100G A122548-10G A122548-1G A122548-250MG A122548-5G
77136-572EA 460.41 USD
77136-572 77136-574 77136-576 77136-578 77136-580
(2-Cyanophenyl)acetic acid 97%
(2-Cyanophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD01646238
CAS Number: 18698-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR