1-Bromo-2-fluoro-4-methoxy-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A121679-100MG A121679-10G A121679-1G A121679-250MG A121679-5G
77136-524EA 6.18 USD
77136-524 77136-526 77136-528 77136-530 77136-532
1-Bromo-2-fluoro-4-methoxy-5-nitrobenzene 97%
1-Bromo-2-fluoro-4-methoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₃
MDL Number: MFCD11847609
CAS Number: 1352244-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR