[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)methanol
A121664-10G
A121664-250MG
77136-520EA
216.43
USD
77136-520
77136-522
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol 97%
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol
Precaución: for Research Use Only
Formula:
C₁₄H₁₁F₃O MW: 252.24 g/mol |
MDL Number:
MFCD01862515 CAS Number: 457889-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...