WYE-354 98%

Supplier: Ambeed
Warning

Synonyms: WYE-354, methyl 4-[6-[4-(methoxycarbonylamino)phenyl]-4-morpholin-4-ylpyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate, Methyl 4-(6-(4-((methoxycarbonyl)amino)phenyl)-4-morpholino-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidine-1-carboxylate, WYE354, WYE 354

A121661-10MG A121661-25MG A121661-5MG A121661-1MG
77136-514EA 82.39 USD
77136-514 77136-516 77136-518 77685-070
WYE-354 98%
WYE-354

Precaución: for Research Use Only

Formula: C₂₄H₂₉N₇O₅
MW: 495.53 g/mol
MDL Number: MFCD18074514
CAS Number: 1062169-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR