(R)-Benzyl 2-amino-3-phenylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Phe-OBzl·HCl

A120451-100G A120451-10G A120451-1G A120451-250MG A120451-25G A120451-5G
77136-464EA 150.38 USD
77136-464 77136-466 77136-468 77136-470 77136-472 77136-474
(R)-Benzyl 2-amino-3-phenylpropanoate hydrochloride 97%
(R)-Benzyl 2-amino-3-phenylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClNO₂
MDL Number: MFCD16038769
CAS Number: 87004-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR