Methyl dihydrojasmonate (cis and trans mixture) 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate, Methyl dihydrojasmonate, Methyl 2-(3-oxo-2-pentylcyclopentyl)acetate, Dihydrojasmonic Acid Methyl Ester

A120330-1000G A120330-100G A120330-25G A120330-500G A120330-10G
77136-456EA 64.72 USD
77136-456 77136-458 77136-460 77136-462 77566-224
Methyl dihydrojasmonate (cis and trans mixture) 95%
Methyl dihydrojasmonate (cis and trans mixture)

Precaución: For research use only.

Formula: C₁₃H₂₂O₃
MW: 226.32 g/mol
MDL Number: MFCD00151188
CAS Number: 24851-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR