(R)-1-Boc-2-Azetidinemethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol, (R)-1-Boc-2-azetidinemethanol
A119262-100MG
A119262-1G
A119262-250MG
A119262-5G
77136-432EA
36.08
USD
77136-432
77136-434
77136-436
77136-438
(R)-1-Boc-2-Azetidinemethanol 98%
(R)-1-Boc-2-Azetidinemethanol
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₃ MW: 187.2362 g/mol |
MDL Number:
MFCD03790937 CAS Number: 161511-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...