(2R,3R,4R,5R)-5-(2-Amino-6-chloro-9H-purin-9-yl)-2-(((4-chlorobenzoyl)oxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl 4-chlorobenzoate 97%

Supplier: Ambeed
Warning

A119192-100MG A119192-1G A119192-250MG
77136-420EA 25.47 USD
77136-420 77136-422 77136-424
(2R,3R,4R,5R)-5-(2-Amino-6-chloro-9H-purin-9-yl)-2-(((4-chlorobenzoyl)oxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl 4-chlorobenzoate 97%
(2R,3R,4R,5R)-5-(2-Amino-6-chloro-9H-purin-9-yl)-2-(((4-chlorobenzoyl)oxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl 4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₂₅H₁₉Cl₃FN₅O₅
MW: 594.81 g/mol
MDL Number: MFCD28501328
CAS Number: 1294481-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR