2-Amino-5-chlorobenzothiazole 97%

Supplier: Ambeed
Warning

A119173-1G A119173-250MG A119173-25G A119173-5G
77136-412EA 7.2 USD
77136-412 77136-414 77136-416 77136-418
2-Amino-5-chlorobenzothiazole 97%
2-Amino-5-chlorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S
MDL Number: MFCD00205355
CAS Number: 20358-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR