1,3,5-Trimethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: O,O,O-1,3,5-Trimethylresorcinol

A118768-1000G A118768-100G A118768-10G A118768-25G A118768-500G A118768-5G
77136-392EA 230.22 USD
77136-392 77136-394 77136-396 77136-398 77136-400 77136-402
1,3,5-Trimethoxybenzene 98%
1,3,5-Trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00008385
CAS Number: 621-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR