(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: (3R)-(+)-1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine, (R)-tert-Butyl (1-benzylpyrrolidin-3-yl)carbamate

A118587-10G A118587-1G A118587-25G A118587-5G
77136-378EA 12.36 USD
77136-378 77136-380 77136-382 77136-384
(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate 95%
(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
MDL Number: MFCD01321148
CAS Number: 131878-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR