trans-3,4-Dimethoxycinnamic acid 95%

Supplier: Ambeed
Warning

A116790-10G A116790-25G A116790-5G A116790-100G A116790-500G
77136-318EA 13.39 USD
77136-318 77136-320 77566-128 77614-918 77685-035
trans-3,4-Dimethoxycinnamic acid 95%
trans-3,4-Dimethoxycinnamic acid

Precaución: For research use only.

Formula: C₁₁H₁₂O₄
MDL Number: MFCD00004387
CAS Number: 14737-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR