(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1R,4S)-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid

A115896-10G A115896-1G A115896-250MG A115896-5G
77136-266EA 157.59 USD
77136-266 77136-268 77136-270 77136-272
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid 95%
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD00211288
CAS Number: 151907-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR