5-Nitroanthranilic acid 98%

Supplier: Ambeed
Warning

A115734-100G A115734-10G A115734-25G A115734-500G A115734-5G A115734-1000G
77136-234EA 46.34 USD
77136-234 77136-236 77136-238 77136-240 77136-242 77745-855
5-Nitroanthranilic acid 98%
5-Nitroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MDL Number: MFCD00017039
CAS Number: 616-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR